Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597612
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597612 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597612 Supplier Invitrogen™ Supplier No. PA597612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CPNE3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a protein which contains two type II C2 domains in the amino-terminus and an A domain-like sequence in the carboxy-terminus. The A domain mediates interactions between integrins and extracellular ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CPNE3
Gene Accession No. O75131
Gene Alias 5430428M23Rik; 5730450C07Rik; C85108; copine 3; copine III; Copine-3; CPN3; Cpne3; KIAA0636; PRO1071
Gene Symbols CPNE3
Host Species Rabbit
Immunogen Recombinant Human Copine-3 protein (297-442AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8895
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.