Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPNE4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585056
Description
Immunogen sequence: AEVPNQVVDY YNGKGIKPKC SSEMYESSRT LAP.
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene is one of several genes that encode a calcium-dependent protein containing two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus.
Specifications
CPNE4 | |
Polyclonal | |
Unconjugated | |
CPNE4 | |
3632411M23Rik; 4933406O10Rik; copine 4; copine 8; copine IV; Copine-4; Copine-8; COPN4; CPN4; Cpne4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96A23 | |
CPNE4 | |
Recombinant protein corresponding to Human CPNE4. Recombinant protein control fragment (Product #RP-108297). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction