Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113260
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113260 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113260 Supplier Invitrogen™ Supplier No. PA5113260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CPNE4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene is one of several genes that encode a calcium-dependent protein containing two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CPNE4
Gene Accession No. Q8BLR2, Q96A23
Gene Alias 3632411M23Rik; 4933406O10Rik; copine 4; copine 8; copine IV; Copine-4; Copine-8; COPN4; CPN4; Cpne4
Gene Symbols CPNE4
Host Species Rabbit
Immunogen Recombinant Human Copine-4 protein (1-280AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131034, 367160, 74020
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.