Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556806 Supplier Invitrogen™ Supplier No. PA556806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DMSDPEMGWV PEPPTMTLGA SRVELRVSCH GLLDRDTLTK PHPCVLLKLY SDEQWVEVER TEVLRSCSSP VFSRVLALEY FFEEKQPLQF HVFDAEDGAT SPRNDTFLGS TECTLGQIVS QTKVTKPLLL KNGKTAGKST ITIVAEEVSG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a brain-specific member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains, and one von Willebrand factor A domain. It may have a role in synaptic plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPNE6
Gene Accession No. O95741
Gene Alias AU067659; BB076446; copine 6; copine VI; copine VI (neuronal); copine-6; Cpne6; MGC137714 protein; N-copine; neuronal copine; neuronal-copine
Gene Symbols CPNE6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPNE6. Recombinant protein control fragment (Product #RP-88929).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9362
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.