Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109583 Supplier Invitrogen™ Supplier No. PA5109583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KYRDKKKNYK SSGTVVLAQC TVEKVHTFLD YIMGGCQISF TVAIDFTASN GDPRSSQSLH CLSPRQPNHY LQALRAVGGI CQDYDSDKRF PAFGFGARIP PNFEVSHDFA INFDPENPEC EEISGVIASY RRCLPQIQLY GPTNVAPIIN RVAEPAQREQ STGQATKYSV LLVLTDGVVS DMAETRTAIV RASRLPMSII IVGVGNADFS DMRLLDGDDG PLRCPRGVPA ARDIVQFVPF RDFKDAAPSA LAKCVLAEVP RQVVEYYASQ GISPGAPRPC TLATTPSPSP.

This gene encodes a brain-specific member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains, and one von Willebrand factor A domain. It may have a role in synaptic plasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CPNE6
Gene Accession No. Q9Z140
Gene Alias AU067659; BB076446; copine 6; copine VI; copine VI (neuronal); copine-6; Cpne6; MGC137714 protein; N-copine; neuronal copine; neuronal-copine
Gene Symbols CPNE6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 268-557 of human CPNE6 (NP_006023.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12891, 691478
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.