Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560337 Supplier Invitrogen™ Supplier No. PA560337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSTTFEEMQK AFEEGQAQWD CVNPKYKQKR RSYKNSGVVV LADLKFHRVY SF Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.

This gene encodes a member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains and one von Willebrand factor A domain. The encoded protein may be involved in membrane trafficking. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cpne7
Gene Accession No. Q9UBL6
Gene Alias AW047065; copine 7; copine 7 protein; copine VII; Copine-7; cpne7; si:ch211-261o3.4
Gene Symbols Cpne7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPNE7. Recombinant protein control fragment (Product #RP-98767).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.