Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558462 Supplier Invitrogen™ Supplier No. PA558462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDSRYNSTAG IGDLNQLSAA IPATRVEVSV SCRNLL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This protein is one of several genes that encode a calcium-dependent protein containing two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPNE8
Gene Accession No. Q86YQ8
Gene Alias 1200003E11Rik; 1500031E20Rik; copine 8; copine 8 protein; copine VIII; Copine-8; Cpne8
Gene Symbols CPNE8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPNE8. Recombinant protein control fragment (Product #RP-97801).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144402
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.