Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543990 Supplier Invitrogen™ Supplier No. PA543990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YDRTVKIDVY DWDRDGSHDF IGEFTTSYRE LSKAQNQFTV YEVLNPRKKC Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

The exact functions of CPNE9 remain unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CPNE9
Gene Accession No. Q8IYJ1
Gene Alias A730016F12Rik; copine family member 9; copine family member IX; copine IX; Copine-9; copine-like protein; Cpne9; mKIAA4217; RGD1309212
Gene Symbols CPNE9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CPNE9.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151835
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.