Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144890 Supplier Invitrogen™ Supplier No. PA5144890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNNNDEDPNC AGIEGVLESY FQ Antigen sequence identity: Mouse 100%; Rat 100%.

The exact functions of CPNE9 remain unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE9
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPNE9
Gene Accession No. Q8IYJ1
Gene Alias A730016F12Rik; copine family member 9; copine family member IX; copine IX; Copine-9; copine-like protein; Cpne9; mKIAA4217; RGD1309212
Gene Symbols CPNE9
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 371-392. Recombinant protein control fragment (Product #RP-109890).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.