Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPO Antibody (OTI1C1), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272386MFV450
Description
CPO Monoclonal antibody specifically detects CPO in Human samples. It is validated for Flow CytometrySpecifications
CPO | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
130749 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1C1 | |
Flow Cytometry | |
carboxypeptidase O, EC 3.4.17, EC 3.4.17.-, MGC138281, MGC138283 | |
Full length human recombinant protein of human CPO(NP_775100) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction