Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPO Monoclonal Antibody (OTI1A1), TrueMAB™, OriGene
SDP

Catalog No. 502540232
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0232 30 μL
50-167-5241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0232 Supplier OriGene Supplier No. TA504589S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CPO Monoclonal antibody specifically detects CPO in Human samples. It is validated for Western Blot

Specifications

Antigen CPO
Applications Western Blot
Classification Monoclonal
Clone OTI1A1
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TAF2
Gene Accession No. Q6P1X5
Gene Alias Carboxypeptidase O; CPO; metallocarboxypeptidase C; metallocarboxypeptidase O
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 782-1121 of human TAF2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6873
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.