Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104426 Supplier Invitrogen™ Supplier No. PA5104426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CPO.

Probable carboxypeptidase which may cleave proteins with C-terminal acidic residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPO
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CPO
Gene Accession No. Q8IVL8
Gene Alias Carboxypeptidase O; CPO; metallocarboxypeptidase C; metallocarboxypeptidase O
Gene Symbols CPO
Host Species Rabbit
Immunogen A synthesized peptide derived from human CPO(Accession Q8IVL8), corresponding to amino acid residues N141-C191.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130749
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.