Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPOX Antibody (36B10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259438H
This item is not returnable.
View return policy
Description
CPOX Monoclonal antibody specifically detects CPOX in Human samples. It is validated for Western Blot, ELISASpecifications
CPOX | |
Monoclonal | |
HRP | |
Coprogen oxidase, coproporphyrinogen oxidase, coproporphyrinogenase, coproporphyrinogen-III oxidase, mitochondrial, COX, CPO, CPXcoproporphyrinogen oxidase (coproporphyria, harderoporphyria), EC 1.3.3.3, HCP | |
Recombinant human CPOX (111-454aa) purified from E. coli | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
1371 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
36B10 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction