Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521635 Supplier Invitrogen™ Supplier No. PA521635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116. Predicted reactivity: Mouse (89%), Rat (88%), Pig (94%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is the sixth enzyme of the heme biosynthetic pathway. The encoded enzyme is soluble and found in the intermembrane space of mitochondria. This enzyme catalyzes the stepwise oxidative decarboxylation of coproporphyrinogen III to protoporphyrinogen IX, a precursor of heme. Defects in this gene are a cause of hereditary coproporphyria (HCP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPOX
Applications Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CPOX
Gene Accession No. P36551
Gene Alias cac; clone 560; coprogen oxidase; Coproporphyrinogen III oxidase, mitochondrial precursor (Coproporphyrinogenase) (Coprogen oxidase) (COX); coproporphyrinogen oxidase; coproporphyrinogenase; coproporphyrinogen-III oxidase, mitochondrial; COX; CPO; Cpox; cpox {ECO:0000312; CPX; EMBL:AAI07665.1}; HCP; nct; oxygen-dependent coproporphyrinogen-III oxidase, mitochondrial
Gene Symbols CPOX
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CPOX. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.