Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576154 Supplier Invitrogen™ Supplier No. PA576154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The protein encoded by this gene is the sixth enzyme of the heme biosynthetic pathway. The encoded enzyme is soluble and found in the intermembrane space of mitochondria. This enzyme catalyzes the stepwise oxidative decarboxylation of coproporphyrinogen III to protoporphyrinogen IX, a precursor of heme. Defects in this gene are a cause of hereditary coproporphyria (HCP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPOX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CPOX
Gene Accession No. P36551, P36552, Q3B7D0
Gene Alias cac; clone 560; coprogen oxidase; Coproporphyrinogen III oxidase, mitochondrial precursor (Coproporphyrinogenase) (Coprogen oxidase) (COX); coproporphyrinogen oxidase; coproporphyrinogenase; coproporphyrinogen-III oxidase, mitochondrial; COX; CPO; Cpox; cpox {ECO:0000312; CPX; EMBL:AAI07665.1}; HCP; nct; oxygen-dependent coproporphyrinogen-III oxidase, mitochondrial
Gene Symbols CPOX
Host Species Rabbit
Immunogen Recombinant fusion protein of human CPOX.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12892, 1371, 304024
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.