Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596257
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA596257 100 μL
1 options

Catalog No. PIPA596257

Supplier: Invitrogen™ PA596257

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSLGRPEEEE DELAHRCSSF MAPPVTDLGE LRRRPGDMKT KMELLILETQ AQVCQALAQV DGGANFSVDR WERKEGGGGI SCVLQDGCVF EKAGVSISVV HGNLSEEAAK QMRSRGKVLK TKDGKLPFCA MGVSSVIHPK NPHAPTIHFN YRYFEVEEAD GNKQWWFGGG CDLTPTYLNQ EDAVHFHRTL KEACDQHGPD LYPKFKKWCD DYFFIAHRGE RRGIGGIFFD DLDSPSKEEV FRFVQSCARA VVPSYIPLVK KHCDDSFTPQ EKLWQQLRRG RYVEFNLLYD RGTKFGLFTP GSRIESILMS LPLTARWEYM HSPSENSKEA EILEVLRHPR DWVR; Positive Samples: HeLa, A375, HepG2, A-549, Mouse liver, Rat liver; Cellular Location: Mitochondrion intermembrane space.

The protein encoded by this gene is the sixth enzyme of the heme biosynthetic pathway. The encoded enzyme is soluble and found in the intermembrane space of mitochondria. This enzyme catalyzes the stepwise oxidative decarboxylation of coproporphyrinogen III to protoporphyrinogen IX, a precursor of heme. Defects in this gene are a cause of hereditary coproporphyria (HCP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPOX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CPOX
Gene Accession No. P36551, P36552, Q3B7D0
Gene Alias cac; clone 560; coprogen oxidase; Coproporphyrinogen III oxidase, mitochondrial precursor (Coproporphyrinogenase) (Coprogen oxidase) (COX); coproporphyrinogen oxidase; coproporphyrinogenase; coproporphyrinogen-III oxidase, mitochondrial; COX; CPO; Cpox; cpox {ECO:0000312; CPX; EMBL:AAI07665.1}; HCP; nct; oxygen-dependent coproporphyrinogen-III oxidase, mitochondrial
Gene Symbols CPOX
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 111-454 of human CPOX (NP_0000883).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12892, 1371, 304024
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.