Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPPED1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565322
Description
Immunogen sequence: DDDYYFNLSKS TRKKLADKFI HAGVRVVFSG HYHRNAGGTY QNLDMVVSSA IGCQLGRDPH GLRVVVVTAE KIVHRYYSLD ELSEKGIEDD LMDL Highest antigen sequence identity to the following orthologs - mouse 77%, rat 80%.
Protein phosphatase that dephosphorylates AKT family kinase specifically at 'Ser-473', blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.
Specifications
CPPED1 | |
Polyclonal | |
Unconjugated | |
CPPED1 | |
C530044N13Rik; calcineurin like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain-containing protein 1; complete S transactivated protein 1; complete S-transactivated protein 1; Cpped1; CSTP1; RGD1306568; Serine/threonine-protein phosphatase CPPED1; Vcalcineurin-like phosphoesterase domain containing 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRF8 | |
CPPED1 | |
Recombinant Human CPPED1. Recombinant protein control fragment (Product #RP-104749). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction