Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPPED1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145455
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145455 200 μL
FEPA5145456 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145455 Supplier Invitrogen™ Supplier No. PA5145455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein phosphatase that dephosphorylates AKT family kinase specifically at 'Ser-473', blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPPED1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CPPED1
Gene Accession No. Q66H71, Q8BFS6, Q9BRF8
Gene Alias C530044N13Rik; calcineurin like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain-containing protein 1; complete S transactivated protein 1; complete S-transactivated protein 1; Cpped1; CSTP1; RGD1306568; Serine/threonine-protein phosphatase CPPED1; Vcalcineurin-like phosphoesterase domain containing 1
Gene Symbols CPPED1
Host Species Rabbit
Immunogen Recombinant Mouse CPPED1 protein, Met1-Glu312 (Accession #Q8BFS6).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223978, 302890, 55313
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.