Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CPPG
GREENER_CHOICESDP

Catalog No. 09725 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-725 5 mg
97-225 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-725 Supplier Tocris Bioscience™ Supplier No. 0972/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent group III mGlu antagonist

CPPG is a potent group II/III mGlu receptor antagonist, with approximately 20-fold selectivity for group III over group II (IC50 values of 2.2 and 46.2 nM respectively). A much less potent antagonist at group I receptors in neonatal rat cortical slices (KB = 0.65 ± 0.07 nM).

Specifications

Chemical Name or Material (RS)-α-Cyclopropyl-4-phosphonophenylglycine
CAS 183364-82-1
Assay Percent Range >98%
Quantity 5 mg
Target Group III mGluR Antagonists
Molecular Formula C11H14NO5P
Formula Weight Observed MW: 271.21
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.