Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPS1 (817-836) Mouse anti-Human, Clone: 5F5, Abnova™

Catalog No. 89933992 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-992 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-992 Supplier Abnova Corporation Supplier No. H00001373M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of CPS1.

The protein encoded by this gene is an enzyme that catalyzes the first committed step of the hepatic urea cycle, which is important in the removal of excess urea from cells. There are two isozymes of this enzyme, and the encoded protein is the mitochondrial form. Three transcript variants encoding different isoforms have been found for this gene. The shortest isoform may not be localized to the mitochondrion. [provided by RefSeq]

Specifications

Antigen CPS1 (817-836)
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 5F5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of CPS1.
Formulation PBS with no preservative; pH 7.4
Gene CPS1
Gene Accession No. NM_001875.4
Gene Symbols CPS1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with BSA) corresponding to internal region of human CPS1.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1373
Test Specificity Antibody Reactive Against Synthetic Peptide.
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.