Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPS1 Recombinant Rabbit Monoclonal Antibody (RM395)
GREENER_CHOICE

Catalog No. PIMA536064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536064 Supplier Invitrogen™ Supplier No. MA536064
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody may also react to mouse or rat CPS1, as predicted by immunogen homology.

The mitochondrial enzyme encoded by this gene catalyzes synthesis of carbamoyl phosphate from ammonia and bicarbonate. This reaction is the first committed step of the urea cycle, which is important in the removal of excess urea from cells. The encoded protein may also represent a core mitochondrial nucleoid protein. Three transcript variants encoding different isoforms have been found for this gene. The shortest isoform may not be localized to the mitochondrion. Mutations in this gene have been associated with carbamoyl phosphate synthetase deficiency, susceptibility to persistent pulmonary hypertension, and susceptibility to venoocclusive disease after bone marrow transplantation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone RM395
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene CPS1
Gene Accession No. P31327
Gene Alias 4732433M03Rik; carbamoyl-phosphate synthase (ammonia); carbamoyl-phosphate synthase [ammonia], mitochondrial; carbamoyl-phosphate synthase 1; carbamoyl-phosphate synthase 1, mitochondrial; carbamoyl-phosphate synthetase 1; carbamoyl-phosphate synthetase 1, mitochondrial; carbamoylphosphate synthetase I; carbamoyl-phosphate synthetase I; carbamyl phosphate synthetase I; carboamyl-phosphate synthetase 1; cps; CPS1; CPSase I; CPSASE1; CPSI; D1Ucla3; PHN
Gene Symbols CPS1
Host Species Rabbit
Immunogen A peptide corresponding to Human CPS1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1373
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.