Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576339 Supplier Thermo Scientific Supplier No. PA576339
Add to Cart
Add to Cart

CPS1 Polyclonal Antibody

CPS1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen CPS1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CPS1
Gene Accession No. P07756, P31327, Q8C196
Gene Alias 4732433M03Rik; carbamoyl-phosphate synthase (ammonia); carbamoyl-phosphate synthase [ammonia], mitochondrial; carbamoyl-phosphate synthase 1; carbamoyl-phosphate synthase 1, mitochondrial; carbamoyl-phosphate synthetase 1; carbamoyl-phosphate synthetase 1, mitochondrial; carbamoylphosphate synthetase I; carbamoyl-phosphate synthetase I; carbamyl phosphate synthetase I; carboamyl-phosphate synthetase 1; cps; CPS1; CPSase I; CPSASE1; CPSI; D1Ucla3; PHN
Gene Symbols CPS1
Host Species Rabbit
Immunogen Recombinant Protein of human CPS1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1373, 227231, 497840
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.