Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109584 Supplier Invitrogen™ Supplier No. PA5109584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKNVVSGKTS ACFEPSLDYM VTKIPRWDLD RFHGTSSRIG SSMKSVGEVM AIGRTFEESF QKALRMCHPS IEGFTPRLPM NKEWPSNLDL RKELSEPSST RIYAIAKAID DNMSLDEIEK LTYIDKWFLY KMRDILNMEK TLKGLNSESM TEETLKRAKE IGFSDKQISK CLGLTEAQTR ELRLKKNIHP WVKQIDTLAA EYPSVTNYLY VTYNGQEHDV N.

The mitochondrial enzyme encoded by this gene catalyzes synthesis of carbamoyl phosphate from ammonia and bicarbonate. This reaction is the first committed step of the urea cycle, which is important in the removal of excess urea from cells. The encoded protein may also represent a core mitochondrial nucleoid protein. Three transcript variants encoding different isoforms have been found for this gene. The shortest isoform may not be localized to the mitochondrion. Mutations in this gene have been associated with carbamoyl phosphate synthetase deficiency, susceptibility to persistent pulmonary hypertension, and susceptibility to venoocclusive disease after bone marrow transplantation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPS1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CPS1
Gene Accession No. P07756, P31327, Q8C196
Gene Alias 4732433M03Rik; carbamoyl-phosphate synthase (ammonia); carbamoyl-phosphate synthase [ammonia], mitochondrial; carbamoyl-phosphate synthase 1; carbamoyl-phosphate synthase 1, mitochondrial; carbamoyl-phosphate synthetase 1; carbamoyl-phosphate synthetase 1, mitochondrial; carbamoylphosphate synthetase I; carbamoyl-phosphate synthetase I; carbamyl phosphate synthetase I; carboamyl-phosphate synthetase 1; cps; CPS1; CPSase I; CPSASE1; CPSI; D1Ucla3; PHN
Gene Symbols CPS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 750-970 of human CPS1 (NP_0018662).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1373, 227231, 497840
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.