Learn More
Description
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-[[5-Amino-1-[(3-methyl-2-thienyl)carbonyl]-1H-1,2,4-triazol-3-yl]amino]-N-[2-[[1-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]ethoxy]ethoxy]ethyl]-1H-1,2,3-triazol-4-yl]methoxy]ethyl]benzenesulfonamide |
| CAS | 2756741-90-7 |
| Quantity | 2 mg |
| Product Line | Tocris |
| Target | Active Degraders |
| Molecular Formula | C38H42N12O10S2 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

