Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPSF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CPSF1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CPSF1 Polyclonal specifically detects CPSF1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CPSF1 | |
Polyclonal | |
Rabbit | |
Human | |
cleavage and polyadenylation specific factor 1, 160kD subunit, cleavage and polyadenylation specific factor 1, 160kDa, Cleavage and polyadenylation specificity factor 160 kDa subunit, cleavage and polyadenylation specificity factor subunit 1, CPSF 160 kDa subunit, CPSF160, HSU37012, P/cl.18, polyadenylation specificity factor | |
CPSF1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
29894 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PHWLLVTGRGALRLHPMAIDGPVDSFAPFHNVNCPRGFLYFNRQGELRISVLPAYLSYDAPWPVRKIPLRCTAHYVAYHVESKVYAVATSTNTPCARIPRMTGEEKEFETIERDERYIHPQQEAFSIQLISPVSWEAIPNARIELQEWE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title