Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPSF1 Polyclonal Antibody

Supplier: Thermo Scientific PA5116722
Description
Specifications
| CPSF1 | |
| Polyclonal | |
| Unconjugated | |
| Cpsf1 | |
| cleavage and polyadenylation specific factor 1; cleavage and polyadenylation specific factor 1, 160kDa; cleavage and polyadenylation specificity factor 1; cleavage and polyadenylation specificity factor 160 kDa subunit; Cleavage and polyadenylation specificity factor subunit 1; CPSF 160 kDa subunit; CPSF1; CPSF160; HSU37012; P/cl.18; polyadenylation specificity factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29894, 94230 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.65 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q10570, Q9EPU4 | |
| Cpsf1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 700-950 of human CPSF1 (NP_0374232). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction