Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPSF100 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen CPSF100
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-0756
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A301583AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine

Component of the cleavage and polyadenylation specificity factor (CPSF) complex that play a key role in pre-mRNA 3'-end formation, recognizing the AAUAAA signal sequence and interacting with poly(A) polymerase and other factors to bring about cleavage and poly(A) addition. Involved in the histone 3' end pre-mRNA processing.
Specifications

Specifications

CPSF100
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
CPSF100, CPSF 100 kDa subunit, CPSF 100kDa subunit, cleavage and polyadenylation specificity factor 100 kDa subunit, cleavage and polyadenylation specificity factor subunit 2
Between 732 and 782
Primary
4° C
CPSF2
Western Blot
Unconjugated
Rabbit
Human, Mouse
O35218, Q9P2I0
51786, 53981
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.