Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117889 Supplier Invitrogen™ Supplier No. PA5117889
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CPSF2 Polyclonal Antibody for Western Blot, IHC (P), IP

CPSF2 gene ontology annotations related to this gene include mRNA cleavage; mRNA polyadenylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CPSF2
Gene Accession No. Q9P2I0
Gene Alias 100kDa; 2610024B04Rik; AI662483; cleavage and polyadenylation specific factor 2; cleavage and polyadenylation specific factor 2, 100kD subunit; cleavage and polyadenylation specific factor 2, 100kDa; cleavage and polyadenylation specificity factor 100 kDa subunit; Cleavage and polyadenylation specificity factor subunit 2; Cpsf; CPSF 100 kDa subunit; CPSF 100kDa subunit; CPSF100; CPSF2; KIAA1367; Mcpsf; mKIAA1367
Gene Symbols CPSF2
Host Species Rabbit
Immunogen E. coli-derived Human CPSF2 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53981
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.