Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557005 Supplier Invitrogen™ Supplier No. PA557005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFNLLCYQLQ KLTGDVEELE IQEKPALKVF KNITVIQEPG MVVLEWLANP SNDMYADTVT TVILEVQSNP KIRKGAVQK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Most eukaryotic mRNA precursors (pre-mRNAs) undergo extensive maturational processing, including cleavage and polyadenylation at the 3-prime end. CPSF3 encodes the 73-kD subunit of the cleavage and polyadenylation specificity factor (CPSF) and is the pre-mRNA 3-prime-end-processing endonuclease (Mandel et al., 2006).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cpsf3
Gene Accession No. Q9QXK7, Q9UKF6
Gene Alias cleavage and polyadenylation specific factor 3; cleavage and polyadenylation specific factor 3, 73kDa; cleavage and polyadenylation specificity factor 3; cleavage and polyadenylation specificity factor 73 kDa subunit; Cleavage and polyadenylation specificity factor subunit 3; CPSF; CPSF 73 kDa subunit; Cpsf3; Cpsf73; CPSF-73; mRNA 3'-end-processing endonuclease CPSF-73; YSH1
Gene Symbols Cpsf3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPSF3. Recombinant protein control fragment (Product #RP-96050).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298916, 51692, 54451
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.