Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPSF3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546133
Description
Peptide sequence: TESTYATTIR DSKRCRERDF LKKVHETVER GGKGAHEPEG AHLLLHGADR Sequence homology: Cow: 100%; Dog: 100%; Goat: 77%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
Chromatin assembly factor I (CAF-I) is required for the assembly of histone octamers onto newly-replicated DNA. CAF-I is composed of three protein subunits, p50, p60, and p150. The protein encoded by this gene corresponds to the p60 subunit and is required for chromatin assembly after replication. The encoded protein is differentially phosphorylated in a cell cycle-dependent manner. In addition, it is normally found in the nucleus except during mitosis, when it is released into the cytoplasm. This protein is a member of the WD-repeat HIR1 family and may also be involved in DNA repair.
Specifications
CPSF3L | |
Polyclonal | |
Unconjugated | |
INTS11 | |
2410006F12Rik; AU044035; cleavage and polyadenylation specific factor 3-like; cleavage and polyadenylation-specific factor 3-like protein; Cpsf3l; CPSF3-like protein; CPSF73L; hypothetical protein LOC553648; int11; Integrator complex subunit 11; integrator complex subunit 11-like; INTS11; Protein related to CPSF subunits of 68 kDa; RC68; RC-68; related to CPSF subunits 68 kDa; RGD1306841; zgc:110671 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54973 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5TA45 | |
INTS11 | |
Synthetic peptide directed towards the C-terminal of human CPSF3L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction