Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118011 Supplier Invitrogen™ Supplier No. PA5118011
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CPSF3L Polyclonal Antibody for IP, Western Blot

Chromatin assembly factor I (CAF-I) is required for the assembly of histone octamers onto newly-replicated DNA. CAF-I is composed of three protein subunits, p50, p60, and p150. The protein encoded by this gene corresponds to the p60 subunit and is required for chromatin assembly after replication. The encoded protein is differentially phosphorylated in a cell cycle-dependent manner. In addition, it is normally found in the nucleus except during mitosis, when it is released into the cytoplasm. This protein is a member of the WD-repeat HIR1 family and may also be involved in DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF3L
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene INTS11
Gene Accession No. Q5TA45
Gene Alias 2410006F12Rik; AU044035; cleavage and polyadenylation specific factor 3-like; cleavage and polyadenylation-specific factor 3-like protein; Cpsf3l; CPSF3-like protein; CPSF73L; hypothetical protein LOC553648; int11; Integrator complex subunit 11; integrator complex subunit 11-like; INTS11; Protein related to CPSF subunits of 68 kDa; RC68; RC-68; related to CPSF subunits 68 kDa; RGD1306841; zgc:110671
Gene Symbols INTS11
Host Species Rabbit
Immunogen E. coli-derived Human INT11/CPSF3L fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54973
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.