Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPSF4 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079826AF700
Description
CPSF4 Polyclonal antibody specifically detects CPSF4 in Human, Mouse samples. It is validated for Western BlotSpecifications
CPSF4 | |
Polyclonal | |
Western Blot | |
30kD, cleavage and polyadenylation specific factor 4, 30kD subunit, cleavage and polyadenylation specific factor 4, 30kDa, CPSF 30 kDa subunit, Neb-1, No arches homolog, no arches-like zinc finger protein, NS1 effector domain-binding protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 220 and 245 of human cleavage and polyadenylation specific factor 4, 30kD subunit using the numbering given in entry NP_006684.1 (GeneID 10898). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
10898 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction