Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561877 Supplier Invitrogen™ Supplier No. PA561877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQEIIASVDH IKFDLEIAVE QQLGAQPLPF PGMDKSGAAV CEFFLKAACG KGGMCPFRHI SGEKT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Inhibition of the nuclear export of poly(A)-containing mRNAs caused by the influenza A virus NS1 protein requires its effector domain. The NS1 effector domain functionally interacts with the cellular 30 kDa subunit of cleavage and polyadenylation specific factor 4, an essential component of the 3' end processing machinery of cellular pre-mRNAs. In influenza virus-infected cells, the NS1 protein is physically associated with cleavage and polyadenylation specific factor 4, 30kD subunit. Binding of the NS1 protein to the 30 kDa protein in vitro prevents CPSF binding to the RNA substrate and inhibits 3' end cleavage and polyadenylation of host pre-mRNAs. Thus the NS1 protein selectively inhibits the nuclear export of cellular, and not viral, mRNAs. Multiple alternatively spliced transcript variants that encode different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPSF4
Gene Accession No. O95639
Gene Alias 30kDa; C79664; cleavage and polyadenylation specific factor 4; cleavage and polyadenylation specific factor 4, 30kD subunit; cleavage and polyadenylation specific factor 4, 30kDa; cleavage and polyadenylation specificity factor 30 kDa subunit; Cleavage and polyadenylation specificity factor subunit 4; clipper homolog; Clipper/CPSF 30K; CPSF 30 kDa subunit; Cpsf30; CPSF4; musculus clipper/cleavage and polyadenylation specificity factor 30 kDa subunit; NAR; NEB1; NEB-1; no; no arches homolog; no arches-like zinc finger protein; NS1 effector domain-binding protein 1
Gene Symbols CPSF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPSF4. Recombinant protein control fragment (Product #RP-100976).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10898
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.