Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595821 Supplier Invitrogen™ Supplier No. PA595821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEVIAGLQG FTFAFELDVE SQKGTGLLPF QGMDKSSSAV CNFFAKGLCV KGMLCPLRHE QGEKLVVCKH WLRGLCRKSD CCDFLHQYDV SKMPVCYFHS KFGNCSNKEC LFLHLKPVLK LQDCPWYNQG FCKEVGPLCK YRHVHQVLCP NYFTGFCPEG PQCQFGHPKM SPPFHPSNVK LQPVNQPWDS TTSTGNTLVS PFQAKPMVHG QRRWSLPQAC SSRAHPAP; Positive Samples: Mouse testis, Mouse brain.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF4L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cpsf4l
Gene Alias 0610010C04Rik; 1500000C01Rik; cleavage and polyadenylation specific factor 4 like; cleavage and polyadenylation specific factor 4-like; Cpsf4l; D11Ertd636e; putative cleavage and polyadenylation specificity factor subunit 4-like protein; RGD1306397
Gene Symbols Cpsf4l
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-228 of mouse Cpsf4l (NP_0809582).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52670
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.