Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPSF4L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983179
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3179 Supplier Bioss Supplier No. BS14037R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CPSF4L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cpsf4l
Gene Accession No. A6NMK7
Gene Alias 0610010C04Rik; 1500000C01Rik; cleavage and polyadenylation specific factor 4 like; cleavage and polyadenylation specific factor 4-like; CPSF4L; D11Ertd636e; Putative cleavage and polyadenylation specificity factor subunit 4-like protein
Gene Symbols Cpsf4l
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CPSF4L .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642843
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.