Learn More
Invitrogen™ CPSF6 Monoclonal Antibody (3F11E1)

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, monkey COS-7 whole cell, rat brain tissue, rat thymus tissue, mouse brain tissue, mouse thymus tissue. IHC: human squamous cell carcinoma tissue, human cervical cancer tissue, human colorectal adenocarcinoma tissue, human ovarian cancer tissue, human spleen tissue, mouse brain tissue, rat brain tissue. ICC/IF: U20S cell. Flow: HepG2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | CPSF6 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3F11E1 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | CPSF6 |
| Gene Accession No. | Q16630, Q6NVF9 |
| Gene Alias | 4733401N12Rik; AI256641; CFIM; CFIM68; cleavage and polyadenylation specific factor 6; cleavage and polyadenylation specific factor 6, 68 kD subunit; cleavage and polyadenylation specific factor 6, 68kDa; cleavage and polyadenylation specificity factor 68 kDa subunit; Cleavage and polyadenylation specificity factor subunit 6; cleavage factor Im complex 68 kDa subunit; CPSF 68 kDa subunit; CPSF6; cpsf6 protein; HPBRII-4; HPBRII-7; pre-mRNA cleavage factor I, 68kD subunit; pre-mRNA cleavage factor Im (68kD); pre-mRNA cleavage factor Im 68 kDa subunit; Protein HPBRII-4/7; Unknown (protein for MGC:142454); wu:fa22f12; zgc:85819 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.