Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103749 Supplier Invitrogen™ Supplier No. PA5103749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CPSF6.

CPSF6 is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors. The cleavage factor complex is composed of four polypeptides. The protein encoded by this gene is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors. The cleavage factor complex is composed of four polypeptides. This gene encodes the 68kD subunit. It has a domain organization reminiscent of spliceosomal proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CPSF6
Gene Accession No. Q16630, Q6NVF9
Gene Alias 4733401N12Rik; AI256641; CFIM; CFIM68; cleavage and polyadenylation specific factor 6; cleavage and polyadenylation specific factor 6, 68 kD subunit; cleavage and polyadenylation specific factor 6, 68kDa; cleavage and polyadenylation specificity factor 68 kDa subunit; Cleavage and polyadenylation specificity factor subunit 6; cleavage factor Im complex 68 kDa subunit; CPSF 68 kDa subunit; CPSF6; cpsf6 protein; HPBRII-4; HPBRII-7; pre-mRNA cleavage factor I, 68kD subunit; pre-mRNA cleavage factor Im (68kD); pre-mRNA cleavage factor Im 68 kDa subunit; Protein HPBRII-4/7; Unknown (protein for MGC:142454); wu:fa22f12; zgc:85819
Gene Symbols CPSF6
Host Species Rabbit
Immunogen A synthesized peptide derived from human CPSF6(Accession Q16630), corresponding to amino acid residues R202-P252.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11052, 432508
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.