Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPSF7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541910
Description
Peptide sequence: GASGSSSRKR HRSRERSPSR SRESSRRHRD LLHNEDRHDD YFQERNREHE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
FLJ12529 is a protein coding gene.
Specifications
CPSF7 | |
Polyclonal | |
Unconjugated | |
CPSF7 | |
5730453I16Rik; AL022757; C330017N18Rik; CFIm59; cleavage and polyadenylation specific factor 7; cleavage and polyadenylation specific factor 7, 59kDa; cleavage and polyadenylation specificity factor 59 kDa subunit; Cleavage and polyadenylation specificity factor subunit 7; cleavage factor Im complex 59 kDa subunit; CPSF 59 kDa subunit; Cpsf7; FLJ12529; LOW QUALITY PROTEIN: cleavage and polyadenylation specificity factor subunit 7; pre mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor Im 59 kDa subunit; RGD1305441; Unknown (protein for IMAGE:7949905) | |
Rabbit | |
Protein A | |
RUO | |
79869 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N684 | |
CPSF7 | |
Synthetic peptide directed towards the C-terminal of human FLJ12529 (aa 411-460). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction