Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559423 Supplier Invitrogen™ Supplier No. PA559423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDDRSSSTEP PPPVRQEPSP KPNNKTPAIL YTYSGLRNRR AAVYVGSFSW WTTDQQLIQV IRSIGVYDVV ELKFAENRA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

FLJ12529 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPSF7
Gene Accession No. Q8N684
Gene Alias 5730453I16Rik; AL022757; C330017N18Rik; CFIm59; cleavage and polyadenylation specific factor 7; cleavage and polyadenylation specific factor 7, 59kDa; cleavage and polyadenylation specificity factor 59 kDa subunit; Cleavage and polyadenylation specificity factor subunit 7; cleavage factor Im complex 59 kDa subunit; CPSF 59 kDa subunit; Cpsf7; FLJ12529; LOW QUALITY PROTEIN: cleavage and polyadenylation specificity factor subunit 7; pre mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor Im 59 kDa subunit; RGD1305441; Unknown (protein for IMAGE:7949905)
Gene Symbols CPSF7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPSF7. Recombinant protein control fragment (Product #RP-97388).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.