Learn More
Invitrogen™ CPSF7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589500
Description
Immunogen sequence: LIDIYADEEF NQDPEFNNTD QIDLYDDVLT ATSQPSDDRS SSTEPPPPVR QEPSPKPNNK TPAILYTYSG LRNRRAAVYV GSFSWWTTDQ QLIQVIRSIG VYDVVELKFA ENRANGQSKG YAEVVVASEN SVHKLLELLP GKVLNGEKVD VRPATRQNLS QFEAQARKRE CVRVPRGGIP PRAHSRDSSD S; Positive Samples: Jurkat, HepG2, Mouse thymus; Cellular Location: Nucleus.
FLJ12529 is a protein coding gene.
Specifications
CPSF7 | |
Polyclonal | |
Unconjugated | |
CPSF7 | |
5730453I16Rik; AL022757; C330017N18Rik; CFIm59; cleavage and polyadenylation specific factor 7; cleavage and polyadenylation specific factor 7, 59kDa; cleavage and polyadenylation specificity factor 59 kDa subunit; Cleavage and polyadenylation specificity factor subunit 7; cleavage factor Im complex 59 kDa subunit; CPSF 59 kDa subunit; Cpsf7; FLJ12529; LOW QUALITY PROTEIN: cleavage and polyadenylation specificity factor subunit 7; pre mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor Im 59 kDa subunit; RGD1305441; Unknown (protein for IMAGE:7949905) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
269061, 79869 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.53 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BTV2, Q8N684 | |
CPSF7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 50-240 of human FLJ12529 (NP_0790873). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.