Learn More
CPSF73 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | CPSF73 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1720
![]() |
Bethyl Laboratories, Inc
IHC00229T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1719
![]() |
Bethyl Laboratories, Inc
IHC00229 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Horse, Rabbit, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Small-eared galago and
Most eukaryotic mRNA precursors (pre-mRNAs) undergo extensive maturational processing, including cleavage and polyadenylation at the 3-prime end. CPSF3 encodes the 73-kD subunit of the cleavage and polyadenylation specificity factor (CPSF) and is the pre-mRNA 3-prime-end-processing endonuclease (Mandel et al., 2006 [PubMed 17128255]).Specifications
CPSF73 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CPSF, CPSF-73, CPSF73, YSH1, CPSF 73 kDa subunit, cleavage and polyadenylation specificity factor 3, cleavage and polyadenylation specificity factor 73 kDa subunit, cleavage and polyadenylation specificity factor subunit 3, mRNA 3′-end-processing endonuclease CPSF-73 | |
Between 634 and 684 | |
Primary | |
4° C | |
CPSF3 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9QXK7, Q9UKF6 | |
51692, 54451 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.