Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPT1A Monoclonal Antibody (5D8G9), CoraLite488, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL48866039100UL
Description
CPT1A Monoclonal antibody specifically detects CPT1A in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
| CPT1A | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| C730027G07; Cpt-1; CPT-Ia; CPT1; CPT1-L; CPT1A; CPTI; L-CPT1 | |
| Protein A | |
| RUO | |
| 1374 | |
| -20° C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Flow Cytometry | |
| 5D8G9 | |
| CoraLite 488 | |
| P50416 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction