Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPT1A Mouse anti-Human, Alexa Fluor™ 647, Clone: 8F6AE9 (also known as 8F6AE9), BD Phrmingen™

Catalog No. BDB569343 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569343 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569343 Supplier BD Biosciences Supplier No. 569343
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CPT1A Monoclonal antibody specifically detects CPT1A in Human samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT1A
Applications Flow Cytometry, Control
Classification Monoclonal
Clone 8F6AE9 (also known as 8F6AE9)
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CPT I; CPT1; CPT1-L; CPT1A; CPTI-L; L-CPT1
Host Species Mouse
Immunogen Human CPT1A (Amino Acids 489-773)
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1374
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.