Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPT1A Mouse anti-Human, Mouse, Rat, Clone: 5D8G9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CPT1A |
---|---|
Clone | 5D8G9 |
Concentration | 1.59 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6479
![]() ![]() |
Proteintech Group Inc
660391IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody can bind the close sequences genes.
The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable), together with a carnitine-acylcarnitine translocase. CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
CPT1A | |
1.59 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CPT I, CPT1, CPT1 L, CPT1A, CPTI L, L CPT1 | |
CPT1A | |
IgG2a | |
Protein A | |
Antibody |
5D8G9 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P32198, P50416, P97742 | |
12894, 1374, 25757 | |
CPT1A Fusion Protein Ag7745 | |
Primary | |
-20°C | |
CPT1A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title