Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPT1A Polyclonal Antibody, CoraLite488, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc CL48815184100UL


Description
CPT1A Polyclonal antibody specifically detects CPT1A in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
CPT1A | |
Polyclonal | |
CoraLite 488 | |
P50416 | |
Rabbit | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Flow Cytometry | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
C730027G07; Cpt-1; CPT-Ia; CPT1; CPT1-L; CPT1A; CPTI; L-CPT1 | |
Antigen affinity chromatography | |
RUO | |
1374 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction