Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPT1B Antibody (43), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290486JF549
Description
CPT1B Monoclonal antibody specifically detects CPT1B in Mouse samples. It is validated for ELISASpecifications
CPT1B | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
43 | |
ELISA | |
carnitine O-palmitoyltransferase 1, muscle isoform, carnitine O-palmitoyltransferase I, mitochondrial muscle isoform, Carnitine O-palmitoyltransferase I, muscle isoform, Carnitine palmitoyltransferase 1B, carnitine palmitoyltransferase 1B (muscle), Carnitine palmitoyltransferase I-like protein, CPT I, CPT1M, CPT1-MMCCPT1, CPTI, CPTI-M, EC 2.3.1, EC 2.3.1.21, FLJ55729, FLJ58750, KIAA1670, MCPT1, M-CPT1 | |
Rabbit immunized with purified, recombinant Mouse CPT1B (Accession#: NP_032596.1, Met 1-Lys 246) | |
0.1 mL | |
Lipid and Metabolism | |
1375 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction