Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPT1B Monoclonal Antibody (OTI2A6)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527328

Catalog No. PIMA527328


Only null left
Add to Cart

Description

Description

CPT1B Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CPT1B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92523
CPT1B
Human recombinant protein fragment corresponding to amino acids 187-440 of CPT1B produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI2A6
Unconjugated
CPT1B
carnitine O-palmitoyltransferase 1, muscle isoform; carnitine O-palmitoyltransferase 1, muscle isoform; synaptonemal complex central element protein 3; carnitine O-palmitoyltransferase 1B; carnitine O-palmitoyltransferase I, mitochondrial muscle isoform; carnitine O-palmitoyltransferase I, muscle isoform; Carnitine palmitoyltransferase 1 beta, muscle isoform; Carnitine palmitoyltransferase 1, muscle; carnitine palmitoyltransferase 1A like; carnitine palmitoyltransferase 1B; carnitine palmitoyltransferase 1B (muscle); carnitine palmitoyltransferase 1B isoform a; carnitine palmitoyltransferase 1b, muscle; carnitine palmitoyltransferase I; carnitine palmitoyltransferase Ibeta 1; carnitine palmitoyltransferase Ibeta 2; carnitine palmitoyltransferase Ibeta 3; carnitine palmitoyltransferase I-like protein; CPT I; Cpt1; cpt1al; CPT1B; CPT1M; Cpt1-m; CPTI; CPTIB; CPT-IB; CPT-Ibeta 1; CPT-Ibeta 3; CPTI-M; EC 2.3.1.21; FLJ55729; FLJ58750; hypothetical protein LOC449677; KIAA1670; LOW QUALITY PROTEIN: carnitine O-palmitoyltransferase 1, muscle isoform; MCCPT1; MCPT1; M-CPT1; M-cpti; muscle-type carnitine palmitoyltransferase I; zgc:103709
Mouse
Affinity Chromatography
RUO
1375
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.