Learn More
CPT1B Mouse anti-Human, Clone: OTI2A6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810597

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and one of the transcripts is bicistronic.Specifications
CPT1B | |
Monoclonal | |
Unconjugated | |
CPT1B | |
CPT1-M, CPT1M, CPTI, CPTI-M, M-CPT1, MCCPT1, MCPT1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1375 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2A6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q92523 | |
CPT1B | |
Human recombit protein fragment corresponding to amino acids 187-440 of human CPT1B produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.