Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPT1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109585 Supplier Invitrogen™ Supplier No. PA5109585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYQVAKALAD DVELYCFQFL PFGKGLIKKC RTSPDAFVQI ALQLAHFRDR GKFCLTYEAS MTRMFREGRT ETVRSCTSES TAFVQAMMEG SHTKADLRDL FQKAAKKHQN MYRLAMTGAG IDRHLFCLYL VSKYLGVSSP FLAEVLSEPW RLSTSQIPQS QIRMFDPEQH PNHLGAGGGF GPVADDGYGV SYMIAGENTI FFHISSKFSS SETNAQRFGN HIRKALLDIA DLFQVPKAYS.

The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene CPT1B
Gene Accession No. Q63704, Q924X2, Q92523
Gene Alias carnitine O-palmitoyltransferase 1, muscle isoform; carnitine O-palmitoyltransferase 1, muscle isoform; synaptonemal complex central element protein 3; carnitine O-palmitoyltransferase 1B; carnitine O-palmitoyltransferase I, mitochondrial muscle isoform; carnitine O-palmitoyltransferase I, muscle isoform; Carnitine palmitoyltransferase 1 beta, muscle isoform; Carnitine palmitoyltransferase 1, muscle; carnitine palmitoyltransferase 1A like; carnitine palmitoyltransferase 1B; carnitine palmitoyltransferase 1B (muscle); carnitine palmitoyltransferase 1B isoform a; carnitine palmitoyltransferase 1b, muscle; carnitine palmitoyltransferase I; carnitine palmitoyltransferase Ibeta 1; carnitine palmitoyltransferase Ibeta 2; carnitine palmitoyltransferase Ibeta 3; carnitine palmitoyltransferase I-like protein; CPT I; Cpt1; cpt1al; CPT1B; CPT1M; Cpt1-m; CPTI; CPTIB; CPT-IB; CPT-Ibeta 1; CPT-Ibeta 3; CPTI-M; EC 2.3.1.21; FLJ55729; FLJ58750; hypothetical protein LOC449677; KIAA1670; LOW QUALITY PROTEIN: carnitine O-palmitoyltransferase 1, muscle isoform; MCCPT1; MCPT1; M-CPT1; M-cpti; muscle-type carnitine palmitoyltransferase I; zgc:103709
Gene Symbols CPT1B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 533-772 of human CPT1B (NP_004368.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12895, 1375, 25756
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.