Learn More
Invitrogen™ CPT1B Recombinant Rabbit Monoclonal Antibody (0R3Z6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557928

Description
Immunogen sequence: FNTTRIPGKD TDVLQHLSDS RHVAVYHKGR FFKLWLYEGA RLLKPQDLEM QFQRILDDPS PPQPGEEKLA ALTAGGRVEW AQARQAFFSS GKNKAALEAI ERAAFFVALD EESYSYDPED EASLSLYGKA LLHGNCYNRW FDKSFTLISF KNGQLGLNAE HAWADAPIIG HLWEFVLGTD SFHLGYTE.
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene.
Specifications
CPT1B | |
Recombinant Monoclonal | |
1.43 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q63704, Q924X2 | |
CPT1B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 313-500 of human CPT1B (NP_004368.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0R3Z6 | |
Unconjugated | |
CPT1B | |
carnitine O-palmitoyltransferase 1, muscle isoform; carnitine O-palmitoyltransferase 1, muscle isoform; synaptonemal complex central element protein 3; carnitine O-palmitoyltransferase 1B; carnitine O-palmitoyltransferase I, mitochondrial muscle isoform; carnitine O-palmitoyltransferase I, muscle isoform; Carnitine palmitoyltransferase 1 beta, muscle isoform; Carnitine palmitoyltransferase 1, muscle; carnitine palmitoyltransferase 1A like; carnitine palmitoyltransferase 1B; carnitine palmitoyltransferase 1B (muscle); carnitine palmitoyltransferase 1B isoform a; carnitine palmitoyltransferase 1b, muscle; carnitine palmitoyltransferase I; carnitine palmitoyltransferase Ibeta 1; carnitine palmitoyltransferase Ibeta 2; carnitine palmitoyltransferase Ibeta 3; carnitine palmitoyltransferase I-like protein; CPT I; Cpt1; cpt1al; CPT1B; CPT1M; Cpt1-m; CPTI; CPTIB; CPT-IB; CPT-Ibeta 1; CPT-Ibeta 3; CPTI-M; EC 2.3.1.21; FLJ55729; FLJ58750; hypothetical protein LOC449677; KIAA1670; LOW QUALITY PROTEIN: carnitine O-palmitoyltransferase 1, muscle isoform; MCCPT1; MCPT1; M-CPT1; M-cpti; muscle-type carnitine palmitoyltransferase I; zgc:103709 | |
Rabbit | |
Protein A | |
RUO | |
12895, 25756 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.